Läs mer
Eltrombopag Olamine, TRC
Produktkod. 30374771
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30374771 25 mg
1 options
Produktkod. 30374771 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE50810025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Eltrombopag Olamine
CAS 496775-62-3
Anteckningar om renhetsgrad HPLC
Molekylformel C25 H22 N4 O4 . 2 C2 H7 N O
Kvantitet 25 mg
Synonym [1,1'-Biphenyl]-3-carboxylic acid, 3'-[(2Z)-2-[1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-, compd. with 2-aminoethanol (1:2), [1,1'-Biphenyl]-3-carboxylic acid, 3'-[(2Z)-[1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazino]-2'-hydroxy-, compd. with 2-aminoethanol (1:2) (9CI), Eltrombopag diethanolamine salt, Eltrombopag olamine, Promacta, Revolade, SB 497115GR, [1,1'-Biphenyl]-3-carboxylic acid, 3'-[(2Z)-2-[1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-, compd. with 2-aminoethanol (1:2), [1,1'-Biphenyl]-3-carboxylic acid, 3'-[(2Z)-[1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazino]-2'-hydroxy-, compd. with 2-aminoethanol (1:2) (9CI), Eltrombopag diethanolamine salt, Eltrombopag olamine, Promacta, Revolade, SB 497115GR, SB-497115GR, SB497115GR Test Mix 1, SB497115GR
InChI formel InChI=1S/C25H22N4O4.2C2H7NO/c1-14-10-11-19(12-15(14)2)29-24(31)22(16(3)28-29)27-26-21-9-5-8-20(23(21)30)17-6-4-7-18(13-17)25(32)33;2*3-1-2-4/h4-13,26,30H,1-3H3,(H,32,33);2*4H,1-3H2/b27-22-;;
LEDER CC1=NN(C(=O)/C/1=N\Nc2cccc(c2O)c3cccc(c3)C(=O)O)c4ccc(C)c(C)c4.NCCO.NCCO
IUPAC-namn 2-aminoethanol;3-[3-[(2Z)-2-[1-(3,4-dimethylphenyl)-3-methyl-5-oxopyrazol-4-ylidene]hydrazinyl]-2-hydroxyphenyl]benzoic acid
Molekylvikt (g/mol) 564.63
Formel vikt 564.2696