Läs mer
Tocris Bioscience™ EMD 281014 hydrochloride
GREENER_CHOICE
Produktkod. 30001548
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001548 10 mg
30001585 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001548 Leverantör Tocris Bioscience™ Leverantörsnummer 4470/10
Selective 5-HT2A antagonist
Selective 5-HT2A antagonist; displays high affinity for human 5-HT2A over 5-HT2C receptors (Ki values are 0.87 and 557 nM respectively). Exhibits low off-target activity (Ki values are >1000 and 1268 nM for human D2 receptors and IKr channels, respectively).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 443144-27-2
Molekylformel C22H22ClFN4O
Specifikationer
Kemiskt namn eller material EMD 281014 hydrochloride
Kvantitet 10 mg
Formel vikt 412.89
Renhet >98%
Molekylvikt (g/mol) 412.893