missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ 25mm Glass Microanalysis Vacuum Filter Holder and Support

Vacuum filter small volumes for particulate or biological contamination analysis

Brand:  EMD Millipore™ XX1002500

Additional Details : Weight : 0.53580kg

 View more versions of this product

Product Code. 10024460

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »


Description

Description

  • Funnel capacity: 15mL
  • Filtration area: 2.5cm2
  • Not autoclavable with filter in place

Specifications

Specifications

Fritted Glass
25mm
15mL
2.5cm2
Videos
View accessory products