Learn More
Merck Hydrosol Stainless Steel Filter Holder and Accessories
Accessories for filter holders for sample preparation.
Brand: Merck XX20047RK
3830.00 SEK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Includes: Lockwheels, wrench set, locking ring, PTFE gasket.
Description
Accessories for filter holders for sample preparation.
The Hydrosol filter holder replacement PartsXX2004704 Funnel and Locking Ring Assembly, stainless steelXX2004702 Base with Screen, gasket and supportXX20047RK Replacement Parts Kit for Hydrosol Filter HolderSpecifications
Not Autoclavable | |
47 mm hydrosol vacuum filter | |
Non-sterile |
Replacement Parts Kit | |
Anodized Aluminum |