Learn More
Merck™ Amicon™ Ultra-0.5 Centrifugal Filter Units
Designed for protein purification, protein concentration and desalting. EMD Millipore Amicon™ Ultra-0.5 Centrifugal Filter Units feature reverse spin capability and are available at two MWCO levels. Constructed with Ultracel membranes, styrene/butadiene housing and polypropylene caps and tubes.
505.00 SEK - 18485.00 SEK
Specifications
Membrane Material Regenerated Cellulose
For Use With (Application) Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
Includes Two microcentrifuge tubes
Length (Metric) 4.99cm
Max. Force 14,000xG for concentration spin; 1000xG for reverse spin
View More Specs
Includes: Two microcentrifuge tubes
Products 20
Product Code Brand MWCO Price Quantity & Availability  
Product Code Brand MWCO Price Quantity & Availability  
10398244 Merck™
UFC5100BK
100,000 d 17345.00 SEK
Pack of 500
Estimated Shipment
15-02-2019
Add to basket
View alternative products »
10527164 Merck™
UFC510008
100,000 d 505.00 SEK
Pack of 8
Estimated Shipment
15-02-2019
Add to basket
View alternative products »
10704645 Merck™
UFC500396
3,000 d 4145.00 SEK
Pack of 96
Sweden Stock
(immediate availability): 1
Add to basket
10795785 Merck™
UFC500308
3,000 d 930.00 SEK
Pack of 8
Estimated Shipment
15-02-2019
Add to basket
View alternative products »
10028713 Merck™
UFC501008
10,000 d 510.00 SEK
Pack of 8
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
10150644 Merck™
UFC5010BK
10,000 d 17345.00 SEK
Pack of 500
Estimated Shipment
15-02-2019
Add to basket
10212584 Merck™
UFC500324
3,000 d 1970.00 SEK
Pack of 24
Sweden Stock
(immediate availability): 3
Add to basket
10130884 Merck™
UFC5003BK
3,000 d 18485.00 SEK
Pack of 500
Estimated Shipment
21-02-2019
Add to basket
10088753 Merck™
UFC501024
10,000 d 2020.00 SEK
Pack of 24
Sweden Stock
(immediate availability): 1
Add to basket
10282244 Merck™
UFC501096
10,000 d 6855.00 SEK
Pack of 96
Sweden Stock
(immediate availability): 1
Add to basket
10328054 Merck™
UFC5030BK
30,000 d 17430.00 SEK
Pack of 500
Estimated Shipment
12-02-2019
Add to basket
View alternative products »
10378434 Merck™
UFC505008
50,000 d 505.00 SEK
Pack of 8
Estimated Shipment
12-02-2019
Add to basket
10257324 Merck™
UFC5050BK
50,000 d 16795.00 SEK
Pack of 500
Estimated Shipment
15-02-2019
Add to basket
View alternative products »
10388244 Merck™
UFC510096
100,000 d 6725.00 SEK
Pack of 96
Estimated Shipment
26-02-2019
Add to basket
10012584 Merck™
UFC503024
30,000 d 2230.00 SEK
Pack of 24
Estimated Shipment
11-02-2019
Add to basket
10505274 Merck™
UFC503008
30,000 d 505.00 SEK
Pack of 8
Estimated Shipment
11-02-2019
Add to basket
10256744 Merck™
UFC503096
30,000 d 6520.00 SEK
Pack of 96
Estimated Shipment
18-02-2019
Add to basket
10194304 Merck™
UFC505024
50,000 d 1985.00 SEK
Pack of 24
Estimated Shipment
08-02-2019
Add to basket
10695445 Merck™
UFC505096
50,000 d 4110.00 SEK
Pack of 96
Estimated Shipment
15-02-2019
Add to basket
View alternative products »
10114694 Merck™
UFC510024
100,000 d 2250.00 SEK
Pack of 24
Estimated Shipment
11-02-2019
Add to basket
Description
Description
  • Concentrates 500μL down to 15μL
  • Concentration factor 25x to 30x
  • Fixed angle rotor: 14,000 × g for concentration spin; 1000 × g for reverse spin
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time - 10 to 30 minutes
  • Reverse spin capability provides consistent recoveries
  • Enables the concentration of proteins with a membrane molecular weight limit of 3 kDa or 10 kDa
Applications:
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting and buffer exchange
Specifications
Specifications
Regenerated Cellulose
Two microcentrifuge tubes
14,000xG for concentration spin; 1000xG for reverse spin
Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
4.99cm
1cm2