Learn More
Merck™ Cartridges and Kits for Milli-Q™ Bowl-Style Water Purification Systems
Pretreatment and purification replacement cartridges
Brand: Merck™ CF0F01205
Additional Details : Weight : 0.50000kg
Description
For Milli-Q™ Bowl systemsSpecifications
Milli-Q™ | |
Milli-Q™ organicfree ultrapure cartridge kit | |
Ion exchange, activated Carbon, depth filtration |
One Super-C™ organics removal cartridge, two ion exchange deionization cartridges, one Organex-Q trace organic removal cartridge, one Millipak™ filter unit | |
Milli-Q™ bowl-style water purification system |