Läs mer
Merck Millipore Filter Holder Vacuum Manifolds
Provide vacuum support for simultaneous filtration of either three or six test samples
Beskrivning
Beskrivning
  • Use Millipore glass, plastic or stainless-steel filter holders fitted with standard No. 8 perforated stoppers over the holder outlets
  • Manifolds connect to single vacuum source and use separate control valves at each station for independent operation
  • Use vent ports on each stainless-steel manifold station to attach Swinnex™ filter holders or Millex™ filter units to maintain sterility on the test filter's downstream side
Stainless-steel Manifolds
  • Stainless-steel body, three-way valves, filter holder supports; anodized aluminum handles; stainless-steel hose connector, pipe plug
  • Units are autoclavable at 121°C (250°F)
PVC Manifold
  • Polyvinylchloride body; stainless-steel two-way valve; polypropylene hose adapters and handles
  • Not autoclavable
Filmer
SDS
Dokument
Dokument
Produktspecifikationer