Läs mer
Merck Millipore Filter Holder Vacuum Manifolds
Provide vacuum support for simultaneous filtration of either three or six test samples
Beskrivning
- Use Millipore glass, plastic or stainless-steel filter holders fitted with standard No. 8 perforated stoppers over the holder outlets
- Manifolds connect to single vacuum source and use separate control valves at each station for independent operation
- Use vent ports on each stainless-steel manifold station to attach Swinnex™ filter holders or Millex™ filter units to maintain sterility on the test filter's downstream side
- Stainless-steel body, three-way valves, filter holder supports; anodized aluminum handles; stainless-steel hose connector, pipe plug
- Units are autoclavable at 121°C (250°F)
- Polyvinylchloride body; stainless-steel two-way valve; polypropylene hose adapters and handles
- Not autoclavable