missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Glass Fiber Filter without Binder

hydrophilic Glass Fiber without Binder Resin filter

442.00 SEK - 835.00 SEK

Specifications

Autoclavable Yes
Color White
For Use With (Application) Used for contamination analysis
Material Borosilicate Microfiber Glass
Filter Type Depth filter
View More Specs

Products 3
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
10498603
View Documents
Merck Millipore
AP4002500
2.5cm 442.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
12-10-2020
Add to basket
 
10498793
View Documents
Merck Millipore
AP4007000
7cm 765.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
12-10-2020
Add to basket
 
10285413
View Documents
Merck Millipore
APFB03700
3.7cm 835.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
16-10-2020
Add to basket
 
Description

Description

hydrophilic Glass Fiber filter with a 0.7μm pore size is a recommended prefilter for 0.9 to 8um filters, especially for heavily contaminated liquids. Can be sterilized by EO, gamma and autoclave.
Specifications

Specifications

Yes
Used for contamination analysis
Depth filter
Plain
White
Borosilicate Microfiber Glass
Sterile
Hydrophilic