Läs mer
MilliporeSigma™ LiChrolut™ Extraction Unit Drying Attachment
Produktkod. 15803971
Change view
Klicka för att se tillgängliga alternativ
Typ:
Drying Attachment, Complete
Förpackningsstorlek:
Varje
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Typ Förpackningsstorlek
15803971 Drying Attachment, Complete Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 15803971 Leverantör MilliporeSigma™ Leverantörsnummer 1198520001
 Begränsad artikel
Beräknad leverans: 17-12-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
for sample preparation
All the individual steps associated with solid-phase extraction can be carried out using the LiChrolut™ extraction unit rapidly and reliability.This transparent and vacuum – suitable unit made of glass can be used to prepare up to 12 samples simultaneously.
TRUSTED_SUSTAINABILITY
Specifikationer
Typ Drying Attachment, Complete