Learn More
Merck™ Microfil™ Filtration System
Convenient filtration system uses presterilized funnels and membranes on lever-action filtration support
8395.00 SEK - 26020.00 SEK
Includes: 47mm gridded MCE membrane filters or EZ-Pak™ membrane cartridge.
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
14842031 View Documents Merck™
MIACFD101
Funnel dispenser; 100mL 8395.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
28-08-2019
Add to basket
10450221 View Documents Merck™
MIAC03P01
Manifold and support; 3-place 26020.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
08-08-2019
Add to basket
10722161 View Documents Merck™
MIAC01P01
Stand alone support This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Disposable, stackable, recyclable polypropylene 100mL or 250mL funnels
  • Eliminates autoclaving and minimizes waste
  • Manifold-mounted supports for laboratory use or stand-alone supports for onsite filtration
  • “Push-fit” funnels seal tightly to manifold supports without clamps or O-rings
  • Leak-free operation and uniform microorganism recoveries
  • Faster than all-glass filter apparatus
  • Lever breaks vacuum and lifts membrane for removal with forceps
Order Info
47mm gridded MCE membrane filters or EZ-Pak™ membrane cartridge
Compliance
CEN, Standard Methods for the Examination of Water and Wastewater, WHO guidelines