Learn More
Merck™ Novagen™ Rosetta™ 2(DE3)pLysS Singles™ Competent Cells
Novagen's Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. Coli.
Brand:  Merck™ 71401-3
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15275187
  • 1870.00 SEK / Each
Estimated Shipment
26-03-2019
Add to basket
Description
Description
Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) on a compatible chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Specifications
Specifications
Guaranteed efficiency: >2 x 106 cfu/μg
11 reactions
Store at ≤-70deg.C
Videos