Learn More
Merck Millipore Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Brand:  Merck Millipore MHA00BSM2
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11983876
  • 819.00 SEK / Pack of 50
Estimated Shipment: 07-09-2021
Add to basket
Description
Description
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifications
Specifications
Orange/Yellow
2-10°C
United States
50 x 2mL
Brettanomyces Selective Broth
Brettanomyces species
For testing wine and beer for Brettanomyces
1 Year
Liquid
Ampule