Learn More
Merck Replacement Solutions for Montage™ BAC96 Miniprep Kit and Montage Plasmid Miniprep96 Kit
Suitable for cell resuspension for plasmid & BAC minipreps
1950.00 SEK
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
11754674
|
Merck
LSKCLS500 |
Suitable for Alkaline Lysis of plasmid and BAC preparation | Cell Lysis Solution |
1950.00 SEK
500mL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11764674
|
Merck
LSKCRS500 |
Plasmid Miniprep, BAC Prep | Cell Resuspension Solution |
1950.00 SEK
500mL |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11704684
|
Merck
LSKNS0500 |
Plasmid Miniprep, BAC Prep | Neutralization Solution |
1950.00 SEK
500mL |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) miniprepsSpecifications
500 mL |