Learn More
Merck Replacement Solutions for Montage™ SEQ96 and SEQ384 Sequencing Reaction Cleanup Kits
provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats.
1965.00 SEK - 2170.00 SEK
Specifications
For Use With (Equipment) | Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits |
---|---|
For Use With (Application) | Plasmid Miniprep, BAC Prep |
Quantity | 500 mL |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11844041
|
Merck
LSKSBW500 |
Wash Solution |
2170.00 SEK
500mL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11724684
|
Merck
LSKSIS500 |
Injection Solution |
1965.00 SEK
500mL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Montage SEQ kits provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats. The unique size-exclusion separation technology eliminates the need for costly gel filtration resins or bind-wash-elute columns.Specifications
Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits | |
Plasmid Miniprep, BAC Prep | |
500 mL |