Learn More
EMD Millipore™ RoClean™ Cleaning Pouches for Millipore Water Purification Systems
Keep your purification system clean with easy-to-use RoClean pouches.
Brand:  EMD Millipore™ ZWACID012
Additional Details : Weight : 0.50000kg Transport : UN number : 1727 Chem class : 1a Pack group : II
 View more versions of this product
Product Code. 10040272
  • 2752.00 SEK / Pack of 12
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
Specifications
Specifications
Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system
Acid cleaning pouch
Videos