Läs mer
MilliporeSigma™ TLC Sprayer and Accessories
Produktkod. 15823881
Change view
Förpackningsstorlek:
Varje
Produktkod. Förpackningsstorlek
15823881 Varje
1 options
Produktkod. 15823881 Leverantör MilliporeSigma™ Leverantörsnummer 1085400001
  • ONLINE EXCLUSIVE PRICE
    11720.00 SEK
    11100.00 SEK / Paket med 1
    Save 620.00 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 16-11-2026
TLC accessories allow professionals to work more quickly, more efficiently, and more reliably
Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.
TRUSTED_SUSTAINABILITY
Specifikationer
Inkluderar 0.8 mm for low viscosity solutions, 1.25 mm optimized for high-viscosity solutions (5 x 0.8 mm bore/1 x 1.25 mm bore)