Läs mer
MilliporeSigma™ TLC Sprayer and Accessories
Produktkod. 15833881
Change view
Klicka för att se tillgängliga alternativ
Förpackningsstorlek:
Paket med 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Förpackningsstorlek
15833881 Paket med 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 15833881 Leverantör MilliporeSigma™ Leverantörsnummer 1085410001
  • ONLINE EXCLUSIVE PRICE
    2170.00 SEK
    2050.00 SEK / Paket med 6
    Save 120.00 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 16-11-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
TLC accessories allow professionals to work more quickly, more efficiently, and more reliably
Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.
TRUSTED_SUSTAINABILITY
Specifikationer
Inkluderar 0.8 mm for low viscosity solutions, 1.25 mm optimized for high-viscosity solutions (5 x 0.8 mm bore/1 x 1.25 mm bore)