Learn More
Merck Adapter, 1/8 NPTM to 1/4
Accessories for filter holders for sample preparation.
Brand: Merck YT3009018
580.00 SEK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Adapts 1/8 or 1/4 in. NPTM fittings to 1/4 in. OD tubingSpecifications
1/4 in. tube O.D. to 1/8 in. NPTM | |
Environmental Test Kits or Supplies | |
0.125 in. |
Water Purification System | |
316 stainless steel | |
0.3175 cm |