missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters

High-recovery membranes for stirred cells

Brand:  EMD Millipore™ 14142

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10034480

  • 4649.00 SEK / Pack of 10


Estimated Shipment
19-07-2018
Add to basket

Description

Description

†43mm nominal diameter.
‡62mm nominal diameter.

Specifications

Specifications

7.6cm
500000 NMWL
Ultrafiltration Disc
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
Videos