missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMILIN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | EMILIN1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837153
|
Proteintech
10643-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16827153
|
Proteintech
10643-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
EMILIN1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y6C2 | |
11117 | |
EMILIN1 Fusion Protein Ag1016 | |
Primary | |
-20°C | |
EMILIN1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EMI, EMILIN, EMILIN 1, EMILIN1, gp115 | |
EMILIN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |