missing translation for 'onlineSavingsMsg'
Learn More

EMILIN1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10643-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827153

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

EMILIN1
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y6C2
EMI, EMILIN, EMILIN 1, EMILIN1, gp115
Rabbit
IgG
150 μL
-20°C
Primary
11117
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EMILIN1
Liquid
EMILIN1
EMILIN1 Fusion Protein Ag1016
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers