Läs mer
Tocris Bioscience™ EML 425
Reversible and non-competitive CBP/p300 inhibitor
Brand: Tocris Bioscience™ 5646/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.Specifikationer
| 5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione | |
| 1675821-32-5 | |
| 50 mg |