missing translation for 'onlineSavingsMsg'
Learn More

EML2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13529-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16871784

  • 4721.67 SEK / 150µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic.
Specifications

Specifications

EML2
Polyclonal
Unconjugated
EML2
ELP70, EMAP 2, EMAP2, EMAPL2, EML2, HuEMAP 2
Rabbit
Antigen Affinity Chromatography
RUO
24139, 72205
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95834, Q7TNG5
Eml2
EML2 Fusion Protein Ag4423
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers