missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EML4 Recombinant Protein
Brand: Abnova™ H00027436-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH08685 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
32.56 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MCYTPCKKYTDMNRQFLEKKEHFFKYLGNTALSDQQGVYLRTSVTFGVAMYNEIYNHDTLRW | |
C2orf2/DKFZp686P18118/ELP120/FLJ10942/FLJ32318/ROPP120 | |
EML4 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
27436 | |
EML4 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EML4 | |
Human | |
Recombinant | |
Solution |