missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emodepside, MedChemExpress
Click to view available options
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
Description
Emodepside (PF 1022-221) is a cyclooctadepsipeptide with broad-spectrum anthelmintic activity.
Specifications
Specifications
| Kemiskt namn eller material | Emodepside |
| Fysisk form | Solid |
| CAS | 155030-63-0 |
| Färg | White |
| Kvantitet | 10 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C60H90N6O14 |
| För användning med (applikation) | COVID-19-immunoregulation |
| Synonym | Bay 44-4400 |
| Löslighetsinformation | DMSO : 120 mg/mL (107.20 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction