missing translation for 'onlineSavingsMsg'
Learn More

EMR3 Antibody (3D7), DyLight 680, Novus Biologicals™

Product Code. 18225670 Shop All Bio Techne Products
Change view
Click to view available options
Kvantitet:
0.1 mL
Unit Size:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
18225670 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18225670 Supplier Novus Biologicals Supplier No. NB10065935FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

EMR3 Monoclonal specifically detects EMR3 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR3
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona 3D7
Konjugera DyLight 680
Utspädning Flow Cytometry
Genaccessionsnr. Q9BY15
Gene Alias egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3
Gensymboler EMR3
Värd art Armenian Hamster
Immunogen ARHO-EMR3-CD97 (EGF1) transfectants.
Reningsmetod Protein G purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin GPCR, Immunology, Innate Immunity
Primär eller sekundär Primary
Gen-ID (Entrez) 84658
Testspecificitet NB100-65935 recognizes human epidermal growth factor (EGF) module-containing mucin-like hormone receptor 3 (EMR3), which is a 56kD member of the EGF-7 transmembrane (TM7) family of adhesion receptors. EMR3 is expressed at the cell surface as a heterodimer. The molecule is predominantly expressed on granulocytes, and at lower levels on mature myeloid cells, monocytes and dendritic cells. EMR3 is absent on lymphocytes, haematopoietic stem cells and myeloid progenitors. Studies suggest that the EMR3 molecule is up-regulated during late stages of neutrophil differentiation and is a marker for terminally differentiated cells. The exact functions of EMR3 and its ligands have not yet been determined.
Målarter Human
Innehåll och lagring Store at 4C in the dark.
Form Purified
Isotyp IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.