Läs mer
Emtricitabine-13C,15N2, TRC
Produktkod. 30134397
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134397 5 mg -
30010256 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134397 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5250025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217820-69-3
Molekylformel 13C C7 H10 F 15N2 N O3 S
Molekylvikt (g/mol) 250.23
Synonym cis-(+/-)-4-Amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone-(2-13C,1,3-15N2), rel-4-Amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone-(2-13C,1,3-15N2), 2',3'-Dideoxy-5-fluoro-3'-thiacytidine-(2-13C,1,3-15N2), 3'-Thia-2',3'-dideoxy-5-fluorocytidine-(2-13C,1,3-15N2), rac-Emtricitabine-(2-13C,1,3-15N2)
IUPAC-namn 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl](2^{13}C,1,3-^{15}N_{2})pyrimidin-2-one
LEDER NC1=[15N][13C](=O)[15N](C=C1F)[C@@H]2CS[C@H](CO)O2
Specifikationer
Kemiskt namn eller material Emtricitabine-13C,15N2
Kvantitet 5 mg
InChI formel InChI=1S/C8H10FN3O3S/c9-4-1-12(8(14)11-7(4)10)5-3-16-6(2-13)15-5/h1,5-6,13H,2-3H2,(H2,10,11,14)/t5-,6+/m0/s1/i8+1,11+1,12+1
Formel vikt 250.0401