Läs mer
Emtricitabine Carboxylic Acid, TRC
Produktkod. 30134986
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134986 100 mg
30003288 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE525035100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1238210-10-0
Molekylformel C8 H8 F N3 O4 S
Molekylvikt (g/mol) 261.23
Synonym 1,3-Oxathiolane-2-carboxylic acid, 5-(4-amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-, (2R,5S)-, (2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic acid, (2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid
IUPAC-namn (2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylic acid
LEDER NC1=NC(=O)N(C=C1F)[C@@H]2CS[C@@H](O2)C(=O)O
Specifikationer
Kemiskt namn eller material Emtricitabine Carboxylic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C8H8FN3O4S/c9-3-1-12(8(15)11-5(3)10)4-2-17-7(16-4)6(13)14/h1,4,7H,2H2,(H,13,14)(H2,10,11,15)/t4-,7+/m0/s1
Formel vikt 261.022