Läs mer
Emtricitabine L-Menthyl Ester ((2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester), Mikromol™
Produktkod. 30458480
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458480 4 x 25 mg
30051482 25 mg
30051524 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30458480 Leverantör Mikromol Leverantörsnummer MM1325.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Emtricitabine L-Menthyl Ester ((2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Emtricitabine L-Menthyl Ester
CAS 764659-72-5
Analyt- eller komponentnamn Emtricitabine L-Menthyl Ester
Frakt skick Room Temperature
Molekylformel C18 H26 F N3 O4 S
Certifikat och efterlevnad ISO 17025
Synonym (2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, Emtricitabine L-Menthyl Ester
InChI formel InChI=1S/C18H26FN3O4S/c1-9(2)11-5-4-10(3)6-13(11)25-16(23)17-26-14(8-27-17)22-7-12(19)15(20)21-18(22)24/h7,9-11,13-14,17H,4-6,8H2,1-3H3,(H2,20,21,24)/t10-,11+,13-,14+,17-/m1/s1
Lösningstyp Neat
LEDER CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@@H]2O[C@@H](CS2)N3C=C(F)C(=NC3=O)N
IUPAC-namn [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylate
Molekylvikt (g/mol) 399.48
Kvantitet 4 x 25 mg
Formel vikt 399.1628 g/mol