missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ EN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514364
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
EN2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the 'engrailed' (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system.
Specifikationer
| EN2 | |
| Polyclonal | |
| Unconjugated | |
| EN2 | |
| AUTS1; BB131122; EN2; En-2; engrailed 2; engrailed homeobox 2; engrailed homolog 2; engrailed-2; homeobox protein en-2; Homeobox protein engrailed-2; hu-En-2; mo-En-2; RGD1561842 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 2020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P19622 | |
| EN2 | |
| KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human EN2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering