Läs mer
Enalapril Diketopiperazine, TRC
Produktkod. 18090830
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18090830 50 mg
18030430 25 mg
18012930 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18090830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55527050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115729-52-7
Molekylformel C20 H26 N2 O4
Molekylvikt (g/mol) 358.43
Synonym Ethyl (2S)-2-[(3S,8aS)-3-Methyl-1,4-dioxooctahydropyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate, Enalapril Maleate Imp. D (EP), Enalapril Diketopiperazine, Pyrrolo[1,2-a]pyrazine-2(1H)-acetic acid, hexahydro-3-methyl-1,4-dioxo-α-(2-phenylethyl)-, ethyl ester, (αS,3S,8aS)-, Pyrrolo[1,2-a]pyrazine-2(1H)-acetic acid, hexahydro-3-methyl-1,4-dioxo-α-(2-phenylethyl)-, ethyl ester, [3S-[2(R*),3α,8aβ]]-, Ethyl (αS,3S,8aS)-hexahydro-3-methyl-1,4-dioxo-α-(2-phenylethyl)pyrrolo[1,2-a]pyrazine-2(1H)-acetate, Enalapril DKP
IUPAC-namn ethyl (2S)-2-[(3S,8aS)-3-methyl-1,4-dioxo-6,7,8,8a-tetrahydro-3H-pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate
LEDER CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)N3CCC[C@H]3C2=O
Specifikationer
Kemiskt namn eller material Enalapril Diketopiperazine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C20H26N2O4/c1-3-26-20(25)17(12-11-15-8-5-4-6-9-15)22-14(2)18(23)21-13-7-10-16(21)19(22)24/h4-6,8-9,14,16-17H,3,7,10-13H2,1-2H3/t14-,16-,17-/m0/s1
Formel vikt 358.1893
Procent renhet >95%