Läs mer
Enalaprilat Dihydrate, TRC
Produktkod. 30041301
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041301 25 mg
18021640 5 mg
18082020 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041301 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55537525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84680-54-6
Synonym Enalaprilat dihydrate∣(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl)pyrrolidine-2-carboxylic acid dehydrate∣Enelapril EP impurity C
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylformel C18H28N2O7
MDL-nummer MFCD00941393
Molekylvikt (g/mol) 384.42, 384.43
InChI-nyckel LRBNLIUVJIKAQR-UHFFFAOYNA-N
LEDER O.O.CC(NC(CCC1=CC=CC=C1)C(O)=O)C(=O)C1CCCN1C(O)=O
IUPAC-namn 2-{2-[(1-carboxy-3-phenylpropyl)amino]propanoyl}pyrrolidine-1-carboxylic acid dihydrate
Produkttyp Metabolite∣Impurity