missing translation for 'onlineSavingsMsg'
Läs mer

Encequidar, MedChemExpress

Produktkod. 16431947
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16431947 50 mg
16426266 100 mg
16408467 5 mg
16405798 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16431947 Leverantör Medchem Express Leverantörsnummer HY13646

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Encequidar (HM30181; HM30181A) is a potent and selective inhibitor of P-glycoprotein.

Encequidar (HM30181

Specifikationer

Kemiskt namn eller material Encequidar
CAS 849675-66-7
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C38H36N6O7
Kvantitet 50 mg
Synonym HM30181 HM30181A
Löslighetsinformation DMSO : 2.4 mg/mL (3.48 mM; Need ultrasonic)
LEDER O=C(C1=CC(C2=CC=CC=C2O1)=O)NC3=CC(OC)=C(OC)C=C3C4=NN(C5=CC=C(CCN6CC7=C(C=C(OC)C(OC)=C7)CC6)C=C5)N=N4
Molekylvikt (g/mol) 688.73
Formel vikt 688.73
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.