Läs mer
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate, TRC
Produktkod. 30469094
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469094 1 g
30465569 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469094 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2886151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate
CAS 1174751-61-1
Molekylformel C9H18N2 . C 2H4O2
Kvantitet 1 g
Synonym USP Granisetron Related Compound E
InChI formel InChI=1S/C9H18N2.C2H4O2/c1-11-8-3-2-4-9(11)6-7(10)5-8;1-2(3)4/h7-9H,2-6,10H2,1H3;1H3,(H,3,4)/t7-,8+,9-;
LEDER N[C@@H]1C[C@@H](N2C)CCC[C@@H]2C1.OC(C)=O
IUPAC-namn (1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine acetate
Molekylvikt (g/mol) 214.3
Formel vikt 214.17