missing translation for 'onlineSavingsMsg'
Läs mer

Endophenazine B, TRC

Produktkod. 30378750
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30378750 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378750 Supplier Toronto Research Chemicals Supplier No. TRCE5554852.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Endophenazine B
CAS 479415-40-2
Molekylformel C19 H18 N2 O3
Kvantitet 2.5 mg
Synonym 1-Phenazinecarboxylic acid, 5,7-dihydro-5-methyl-9-(3-methyl-2-buten-1-yl)-7-oxo-, 1-Phenazinecarboxylic acid, 5,7-dihydro-5-methyl-9-(3-methyl-2-butenyl)-7-oxo- (9CI), 5,7-Dihydro-5-methyl-9-(3-methyl-2-buten-1-yl)-7-oxo-1-phenazinecarboxylic acid, Endophenazine B
InChI formel InChI=1S/C19H18N2O3/c1-11(2)7-8-12-9-13(22)10-16-17(12)20-18-14(19(23)24)5-4-6-15(18)21(16)3/h4-7,9-10H,8H2,1-3H3,(H,23,24)
LEDER CN1C2=CC(=O)C=C(CC=C(C)C)C2=Nc3c1cccc3C(=O)O
IUPAC-namn 5-methyl-9-(3-methylbut-2-enyl)-7-oxophenazine-1-carboxylic acid
Molekylvikt (g/mol) 322.36
Formel vikt 322.1317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.