Läs mer
Endovion, MedChemExpress
Produktkod. 16482936
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16482936 25 mg 25mg
16470417 50 mg -
16426767 5 mg -
16415046 100 mg -
16404068 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16482936 Leverantör Medchem Express Leverantörsnummer HY105917
MedChemExpress Endovion (NS3728) is a pharmacological anion channel inhibitor (like chloride channel) and the specific VRAC/VSOAC blocker. Endovion (NS3728) is also an Anoctamin-1 (ANO 1) channel inhibitor.
Endovion (NS3728) is a pharmacological anion channel inhibitor (like chloride channel) and the specific VRAC/VSOAC blocker. Endovion (NS3728) is also an Anoctamin-1 (ANO 1) channel inhibitor.
Specifikationer
Kemiskt namn eller material Endovion
CAS 265646-85-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C16H9BrF6N6O
Kvantitet 25 mg
Synonym NS3728
Löslighetsinformation DMSO : 125 mg/mL (252.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER FC(F)(F)C1=CC(C(F)(F)F)=CC(NC(NC2=C(C3=NN=NN3)C=C(Br)C=C2)=O)=C1
Molekylvikt (g/mol) 495.18
Formel vikt 495.18
Procent renhet 99.13%
Kvalitet Research
Fysisk form Solid