Läs mer
Azpack™ Engraver
Produktkod. 12329399
Change view
Kvantitet:
1 st
Förpackningsstorlek:
Varje
Produktkod. Kvantitet Förpackningsstorlek
12329399 1 st Varje
1 options
Produktkod. 12329399 Leverantör Azpack™ Leverantörsnummer 12329399
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Mark or decorate metal, stone, plastic, glass, porcelain. Azpack™ Engraver works according to the vibration principle using AC frequency.
  • Intensity can be infinitely adjusted
  • Includes adapter and steel tip.
  • 1.75 metre ext cord.
  • Power supply: 220 V, 50 Hz, 25 Watts
  • Optional tips available separately
Specifikationer
Inkluderar Adapter and steel tip
Antal per förpackning 1 st.
Kvantitet 1 st