Läs mer
Eniluracil, MedChemExpress
Produktkod. 16450387
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16450387 50 mg
16404038 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16450387 Leverantör Medchem Express Leverantörsnummer HY10533
MedChemExpress Eniluracil (5-Ethynyluracil), a uracil analogue and a mechanism-based irreversible inhibitor of dihydropyrimidine dehydrogenase (DPD), increases the oral bioavailability of 5-fluorouracil (5-FU) to 100%, facilitating uniform absorption and predictable toxicity.
Eniluracil (5-Ethynyluracil), a uracil analogue and a mechanism-based irreversible inhibitor of dihydropyrimidine dehydrogenase (DPD), increases the oral bioavailability of 5-fluorouracil (5-FU) to 100%, facilitating uniform absorption and predictable toxicity.
Specifikationer
Kemiskt namn eller material Eniluracil
Fysisk form Powder
CAS 59989-18-3
Färg Off-White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C6H4N2O2
För användning med (applikation) Neuroscience-Neuromodulation
Synonym 5-Ethynyluracil GW776C85
Löslighetsinformation DMSO : 41.67 mg/mL (306.15 mM; Need ultrasonic)
LEDER O=C(N1)NC=C(C#C)C1=O
Molekylvikt (g/mol) 136.11
Formel vikt 136.11
Procent renhet 98.0%