Läs mer
ENMD-2076 Tartrate, MedChemExpress
Produktkod. 16494328
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16494328 10 mg
16467047 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16494328 Leverantör Medchem Express Leverantörsnummer HY10987
MedChemExpress ENMD-2076 Tartrate is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively.
ENMD-2076 Tartrate is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively.
Specifikationer
Kemiskt namn eller material ENMD-2076 Tartrate
Fysisk form Solid
CAS 1291074-87-7
Färg Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H31N7O6
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 25 mg/mL (47.57 mM; Need ultrasonic)
LEDER O[C@H](C(O)=O)[C@@H](C(O)=O)O.CC1=CC(NC2=NC(/C=C/C3=CC=CC=C3)=NC(N4CCN(CC4)C)=C2)=NN1
Molekylvikt (g/mol) 525.56
Formel vikt 525.56
Procent renhet 98.59%