missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOX1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23750-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
ENOX1 | |
Polyclonal | |
Unconjugated | |
ENOX1 | |
cCNOX, CNOX, Constitutive Ecto NOX, ENOX1, PIG38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55068 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TC92 | |
ENOX1 | |
ENOX1 Fusion Protein Ag20715 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |