Learn More
Enoxacin, TRC
Product Code. 30008728
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008728 500 mg 500mg
30134407 5 g 5g
30133433 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008728 Supplier Toronto Research Chemicals Supplier No. TRCE557600500MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74011-58-8
Molecular Formula C15 H17 F N4 O3
Molecular Weight (g/mol) 320.32
Synonym 1,8-Naphthyridine-3-carboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)- (9CI, ACI), 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid (ACI), 1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-6-fluoro-4-oxo-7-piperazin-4-ium-1-yl-1,8-naphthyridine-3-carboxylate, AT 2266, CI 919, Enofloxacin, Enofloxacine, Enoksetin, Enoxacine, ENOXOR, Flumark, MeSH ID: D015365, NSC 629661, PD 107779, Penetrex, Penetrex (pharmaceutical)
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(nc12)N3CCNCC3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C15H17FN4O3/c1-2-19-8-10(15(22)23)12(21)9-7-11(16)14(18-13(9)19)20-5-3-17-4-6-20/h7-8,17H,2-6H2,1H3,(H,22,23)
Formula Weight 320.1285
Percent Purity >95%
Chemical Name or Material Enoxacin
Grade HPLC