Läs mer
Enpiroline, TRC
Produktkod. 30370340
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370340 10 mg
30383955 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30370340 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55765010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66364-73-6
Molekylformel C19 H18 F6 N2 O
Molekylvikt (g/mol) 404.35
Synonym αr)-Rel-α-(2R)-2-Piperidinyl-2-(Trifluoromethyl)-6-[4-(Trifluoromethyl)Phenyl]-4-Pyridinemethanol, (R*,R*)-(±)-α-2-Piperidinyl-2-(Trifluoromethyl)-6-[4-(Trifluoromethyl)Phenyl]-4-Pyridinemethanol
LEDER O[C@@H]([C@H]1Ccccn1)C2Cc(Nc(C2)C(F)(F)F)C3Ccc(Cc3)C(F)(F)F
Specifikationer
Kemiskt namn eller material Enpiroline
Kvantitet 10 mg
InChI formel InChI=1S/C19H18F6N2O/c20-18(21,22)13-6-4-11(5-7-13)15-9-12(10-16(27-15)19(23,24)25)17(28)14-3-1-2-8-26-14/h4-7,9-10,14,17,26,28H,1-3,8H2
Formel vikt 404.1323