missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ENSA (Human) Recombinant Protein
Human ENSA full-length ORF ( AAH00436, 1 a.a. - 121 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002029-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSQKQEEENPAEETGEEKQDTQEKEGILPERAEEAKLKAKYPSLGQKPGGSDFLMKRLQKGQKYFDSGDYNMAKAKMKNKQLPSAGPDKNLVTGDHIPTPQDLPQRKSSLVTSKLAGGQVE
Specifications
AAH00436 | |
endosulfine alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENSA | |
GST |
2029 | |
Wheat germ expression system | |
25 μg | |
MGC4319, MGC78563, MGC8394 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |