Läs mer
Enstar, TRC
Produktkod. 17991305
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991305 25 mg
17961725 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17991305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55790325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42588-37-4
Molekylformel C18 H28 O2
Molekylvikt (g/mol) 276.41
Synonym 2,4-Dodecadienoic acid, 3,7,11-trimethyl-, 2-propynyl ester, (2E,4E)- (9CI), 2,4-Dodecadienoic acid, 3,7,11-trimethyl-, 2-propynyl ester, (E,E)-, ENT 70531, Enstar, Kinoprene, XR 777, ZR 77
IUPAC-namn prop-2-ynyl (2E,4E)-3,7,11-trimethyldodeca-2,4-dienoate
LEDER CC(C)CCCC(C)C\C=C\C(=C\C(=O)OCC#C)\C
Specifikationer
Kemiskt namn eller material Kinoprene
Kvantitet 25 mg
InChI formel InChI=1S/C18H28O2/c1-6-13-20-18(19)14-17(5)12-8-11-16(4)10-7-9-15(2)3/h1,8,12,14-16H,7,9-11,13H2,2-5H3/b12-8+,17-14+
Formel vikt 276.2089