Läs mer
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol™
Produktkod. 17911056
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17911056 25 mg
17456683 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17911056 Leverantör Mikromol Leverantörsnummer MM0033.020025
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
ent-3-Methoxy-17-methylmorphinan-10-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material ent-3-Methoxy-17-methylmorphinan-10-one (10-Ketodextromethorphan)
CAS 57969-05-8
Analyt- eller komponentnamn ent-3-Methoxy-17-methylmorphinan-10-one (10-Ketodextromethorphan)
Frakt skick Room Temperature
Molekylformel C18 H23 N O2
Certifikat och efterlevnad ISO 17025
Synonym ent-3-Methoxy-17-methylmorphinan-10-one, 10-Keto Dextromethorphan, Dextromethorphan Hydrobromide Imp. C (EP), Dextromethorphan Imp. C (EP), Dextromethorphan Related Compound C, Dextromethorphan Hydrobromide Impurity C, Dextromethorphan Impurity C
InChI formel InChI=1S/C18H23NO2/c1-19-10-9-18-8-4-3-5-14(18)16(19)17(20)13-7-6-12(21-2)11-15(13)18/h6-7,11,14,16H,3-5,8-10H2,1-2H3/t14-,16-,18+/m1/s1
LEDER COc1ccc2C(=O)[C@H]3[C@H]4CCCC[C@@]4(CCN3C)c2c1
IUPAC-namn (9α,13α,14α)-3-Methoxy-17-methylmorphinan-10-one
Molekylvikt (g/mol) 285.38
Kvantitet 25 mg
Formel vikt 285.1729 g/mol