Läs mer
Ent-Alpha-C Dapagliflozin (Dapagliflozin Impurity), TRC
Produktkod. 30357599
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357599 5 mg
30352505 10 mg
30349223 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30357599 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1853655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2452300-81-9
Molekylformel C21H25ClO6
Molekylvikt (g/mol) 408.87
Synonym (2S,3S,4S,5R,6S)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
IUPAC-namn (2S,3S,4S,5R,6S)-2-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-6-(hydroxymethyl)oxane-3,4,5-triol
LEDER CCOc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]3O)cc1
Specifikationer
Kemiskt namn eller material Ent-alpha-C Dapagliflozin (Dapagliflozin Impurity)
Kvantitet 5 mg
InChI formel InChI=1S/C21H25ClO6/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21-/m0/s1
Formel vikt 408.134