Läs mer
ent-Cinacalcet Hydrochloride, TRC
Produktkod. 30368236
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368236 10 mg
30363284 5 mg
30368069 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC44180510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217809-88-5
Molekylformel C22 H22 F3 N . Cl H
Molekylvikt (g/mol) 393.87
Synonym 1-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (αS)-, (S)-Cinacalcet hydrochloride, ent-Cinacalcet hydrochloride, [(1S)-1-(Naphthalen-1-yl)ethyl]([3-[3-(trifluoromethyl)phenyl]propyl])amine hydrochloride
IUPAC-namn N-[(1S)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride
LEDER Cl.C[C@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
Specifikationer
Kemiskt namn eller material ent-Cinacalcet Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m0./s1
Formel vikt 393.1471