Läs mer
ent-Levobunolol Hydrochloride (R-Bunolol Hydrochloride), TRC
Produktkod. 30137358
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137358 1 g
30005427 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137358 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3350101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27867-05-6
Molekylformel C17 H25 N O3 . Cl H
Molekylvikt (g/mol) 327.85
Synonym 1(2H)-Naphthalenone, 5-[(2R)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (1:1), 1(2H)-Naphthalenone, 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (+)- (8CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (R)- (9CI), (+)-Bunolol hydrochloride, d-Bunolol hydrochloride, (R)-Bunolol hydrochloride
IUPAC-namn 5-[(2R)-3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride
LEDER Cl.CC(C)(C)NC[C@@H](O)COc1cccc2C(=O)CCCc12
Specifikationer
Kemiskt namn eller material ent-Levobunolol, Hydrochloride
Kvantitet 1 g
InChI formel InChI=1S/C17H25NO3.ClH/c1-17(2,3)18-10-12(19)11-21-16-9-5-6-13-14(16)7-4-8-15(13)20;/h5-6,9,12,18-19H,4,7-8,10-11H2,1-3H3;1H/t12-;/m1./s1
Formel vikt 327.1601