Läs mer
Tocris Bioscience™ ent-LP 99
GREENER_CHOICE
Produktkod. 18707136
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18707136 25 mg
18746236 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18707136 Leverantör Tocris Bioscience™ Leverantörsnummer 5497/25
Negative Control for LP 99
ent-LP 99 is a negative Control for LP 99 (Cat.No. 5496).
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[(2S,3R)-2-(4-Chlorophenyl)-1-(1,2-dihydro-1,4-dimethyl-2-oxo-7-quinolinyl)-6-oxo-3-piperidinyl]-2-methyl-1-propanesulfonamide
CAS 1808948-28-8
Kvantitet 25 mg
Molekylformel C26H30ClN3O4S
Formel vikt 516.05
Renhet 98%