Läs mer
Ent-Mavacamten, TRC
Produktkod. 30475256
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475256 50 mg
30469840 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475256 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29982050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ent-Mavacamten
CAS 1642288-64-9
Molekylformel C15H19N3O2
Kvantitet 50 mg
Synonym Mavacamten Impurity
InChI formel InChI=1S/C15H19N3O2/c1-10(2)18-14(19)9-13(17-15(18)20)16-11(3)12-7-5-4-6-8-12/h4-11,16H,1-3H3,(H,17,20)/t11-/m1/s1
LEDER O=C1NC(N[C@H](C)C2=CC=CC=C2)=CC(N1C(C)C)=O
IUPAC-namn (R)-3-isopropyl-6-((1-phenylethyl)amino)pyrimidine-2,4(1H,3H)-dione
Molekylvikt (g/mol) 273.34
Formel vikt 273.15